Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009700.2 | NP_033830.2 | 2 | 349 | ||
GenBank mRNA | AF469168.1 | - | 2 | 423 | ||
AF469169.1 | - | 2 | 605 | |||
AK039777.1 | - | 2 | 351 | |||
AK044074.1 | - | 2 | 288 | |||
AY484964.1 | - | 2 | 355 | |||
AY484965.1 | - | 3 | 539 | |||
AY484966.1 | - | 3 | 485 | |||
AY728043.1 | - | 2 | 369 | |||
AY728044.1 | - | 3 | 553 | |||
AY728045.1 | - | 2 | 481 | |||
AY803942.1 | - | 3 | 460 | |||
AY803943.1 | - | 4 | 590 | |||
AY803944.1 | - | 5 | 774 | |||
BB628835.1 | - | 2 | 350 | |||
BC024526.1 | - | 2 | 295 | 4985265 | ||
CD803766.1 | - | 2 | 474 | |||
CT010362.1 | - | 1 | 296 | |||
U88623.1 | - | 2 | 350 |