Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009700.2 | NP_033830.2 | 3 | 648 | ||
GenBank mRNA | AF469168.1 | - | 3 | 722 | ||
AF469169.1 | - | 3 | 904 | |||
AK039777.1 | - | 3 | 651 | |||
AK044074.1 | - | 3 | 587 | |||
AK045357.1 | - | 2 | 299 | |||
AY484964.1 | - | 3 | 654 | |||
AY484965.1 | - | 4 | 838 | |||
AY484966.1 | - | 4 | 784 | |||
AY728043.1 | - | 3 | 668 | |||
AY728044.1 | - | 4 | 852 | |||
AY728045.1 | - | 3 | 780 | |||
AY803942.1 | - | 4 | 759 | |||
AY803943.1 | - | 5 | 889 | |||
AY803944.1 | - | 6 | 1073 | |||
BC024526.1 | - | 3 | 594 | 4985265 | ||
CT010362.1 | - | 2 | 595 | |||
U33012.1 | - | 2 | 610 | |||
U48397.1 | - | 2 | 730 | |||
U48398.1 | - | 2 | 601 | |||
U48399.1 | - | 4 | 712 | |||
U88623.1 | - | 3 | 649 |