Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009700.2 | NP_033830.2 | 4 | 720 | ||
GenBank mRNA | AF469168.1 | - | 4 | 794 | ||
AF469169.1 | - | 4 | 976 | |||
AK039777.1 | - | 4 | 723 | |||
AK044074.1 | - | 4 | 659 | |||
AK045357.1 | - | 3 | 371 | |||
AY484964.1 | - | 4 | 726 | |||
AY484965.1 | - | 5 | 910 | |||
AY484966.1 | - | 5 | 856 | |||
AY728043.1 | - | 4 | 740 | |||
AY728044.1 | - | 5 | 924 | |||
AY728045.1 | - | 4 | 852 | |||
AY803942.1 | - | 5 | 831 | |||
AY803943.1 | - | 6 | 961 | |||
AY803944.1 | - | 7 | 1145 | |||
BC024526.1 | - | 4 | 666 | 4985265 | ||
CT010362.1 | - | 3 | 667 | |||
U33012.1 | - | 3 | 682 | |||
U48397.1 | - | 3 | 802 | |||
U48398.1 | - | 3 | 673 | |||
U48399.1 | - | 5 | 784 | |||
U88623.1 | - | 4 | 721 |