Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009701.4 | NP_033831.1 | 3 | 830 | ||
GenBank mRNA | AF087654.1 | - | 3 | 1079 | ||
AK004736.1 | - | 3 | 708 | |||
AK009302.1 | - | 3 | 715 | |||
BC026379.1 | - | 2 | 1024 | |||
BC150769.1 | - | 4 | 876 | 9087680 |