Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007473.4 | NP_031499.1 | 5 | 782 | ||
GenBank mRNA | AB010100.1 | - | 5 | 673 | ||
AB056091.1 | - | 5 | 616 | |||
AK076642.1 | - | 5 | 750 | |||
AK131744.1 | - | 5 | 750 | |||
BC022223.1 | - | 3 | 412 | 4242417 |