Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109045.1 | NP_001102515.1 | 2 | 316 | ||
NM_007474.2 | NP_031500.1 | 2 | 319 | |||
GenBank mRNA | AF018952.1 | - | 2 | 301 | ||
BC010982.1 | - | 2 | 261 | 4224850 | ||
BY713653.1 | - | 2 | 321 | |||
CF617211.1 | - | 2 | 222 |