Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109045.1 | NP_001102515.1 | 5, 6 | 895 | ||
NM_007474.2 | NP_031500.1 | 5, 6 | 898 | |||
GenBank mRNA | AF018952.1 | - | 5, 6 | 880 | ||
AK146660.1 | - | 4, 5 | 577 | |||
BC010982.1 | - | 5, 6 | 840 | 4224850 |