Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Areg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009704.3 | NP_033834.1 | 4 | 842 | ||
GenBank mRNA | AK018590.1 | - | 4 | 858 | ||
AK020273.1 | - | 4 | 858 | |||
BC009138.1 | - | 4 | 842 | 3597695 | ||
CT010320.1 | - | 4 | 643 | |||
D12648.1 | - | 4 | 669 |