Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031259.1 | - | 2 | 170 | ||
AK042977.1 | - | 2 | 210 | |||
AK053123.1 | - | 2 | 628 | |||
AK077591.1 | - | 2 | 183 | |||
AK167133.1 | - | 2 | 180 | |||
AK167601.1 | - | 2 | 184 | |||
AK167653.1 | - | 2 | 183 | |||
AK169094.1 | - | 2 | 180 | |||
AK169412.1 | - | 2 | 180 | |||
BC010487.1 | - | 2 | 463 | 3483748 | ||
BY129623.1 | - | 2 | 222 | |||
D87899.1 | - | 2 | 175 |