Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007482.3 | NP_031508.1 | 2, 3 | 234 | ||
GenBank mRNA | AK149471.1 | - | 2, 3 | 217 | ||
AK171417.1 | - | 1, 2 | 68 | |||
BC013341.1 | - | 2, 3 | 211 | 4163626 | ||
BC050005.1 | - | 2, 3 | 248 | 3990246 | ||
CB947842.1 | - | 2, 3 | 238 | |||
CT010173.1 | - | 2, 3 | 132 | |||
U51805.1 | - | 2, 3 | 273 |