Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287530.1 | NP_001274459.1 | 11 | 1933 | ||
NM_009707.4 | NP_033837.2 | 11 | 3301 | |||
NM_178754.3 | NP_848869.1 | 11 | 1844 | |||
GenBank mRNA | AF012273.2 | - | 2423 | |||
AF177664.1 | - | 2562 | ||||
AK048162.1 | - | 11 | 1798 | |||
AK048507.1 | - | 11 | 1799 | |||
AK077996.1 | - | 11 | 1806 | |||
AK079651.1 | - | 13 | 1896 | |||
AK166015.1 | - | 11 | 1764 |