Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgdib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301301.1 | NP_001288230.1 | 4 | 391 | ||
NM_001301303.1 | NP_001288232.1 | 4 | 378 | |||
NM_001301305.1 | NP_001288234.1 | 4 | 370 | |||
NM_001301308.1 | NP_001288237.1 | 3 | 220 | |||
NM_001301309.1 | NP_001288238.1 | 3 | 201 | |||
NM_007486.5 | NP_031512.1 | 4 | 410 | |||
GenBank mRNA | AA219822.1 | - | 4 | 344 | ||
AK002516.1 | - | 4 | 372 | |||
AK169733.1 | - | 4 | 368 | |||
BB755757.1 | - | 4 | 360 | |||
BC031763.1 | - | 4 | 323 | 4923219 | ||
L07918.1 | - | 4 | 324 |