Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phox2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008887.2 | NP_032913.1 | 2 | 486 | ||
GenBank mRNA | CJ133172.1 | - | 2 | 236 | ||
X75014.1 | - | 2 | 328 |