Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037737.2 | NP_001032826.1 | 5, 6 | 428 | ||
NM_009709.4 | NP_033839.2 | 4, 5 | 383 | |||
GenBank mRNA | AK028546.1 | - | 4, 5 | 370 | ||
AK040475.1 | - | 5, 6 | 430 | |||
AK141362.1 | - | 5, 6 | 299 | |||
AK145415.1 | - | 4, 5 | 338 | |||
AK152473.1 | - | 5, 6 | 410 | |||
AK153355.1 | - | 5, 6 | 410 | |||
AK220458.1 | - | 1, 2 | 559 | |||
BC012870.1 | - | 5, 6 | 390 | 3964932 | ||
U10325.1 | - | 5, 6 | 292 |