Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Art5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291354.1 | NP_001278283.1 | 3 | 595 | ||
NM_007491.2 | NP_031517.2 | 3 | 647 | |||
GenBank mRNA | AJ295843.1 | - | 2 | 98 | ||
AJ297547.1 | - | 2 | 98 | |||
AJ297548.1 | - | 2 | 98 | |||
AK132854.1 | - | 3 | 648 | |||
BC116823.1 | - | 3 | 459 | 40126142 | ||
BC116849.1 | - | 3 | 459 | 40126168 | ||
BC144825.1 | - | 3 | 459 | 9053373 | ||
BC144826.1 | - | 3 | 407 | 9053374 | ||
BC144827.1 | - | 2 | 646 | 9053375 | ||
BC144836.1 | - | 3 | 407 | |||
U60881.1 | - | 480 |