Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asgr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291131.1 | NP_001278060.1 | 5, 6 | 402 | ||
NM_009714.3 | NP_033844.1 | 5, 6 | 512 | |||
GenBank mRNA | AK132959.1 | - | 5, 6 | 403 | ||
BC014779.1 | - | 5, 6 | 418 | |||
BC022106.1 | - | 5, 6 | 474 | 5102469 | ||
CB948939.1 | - | 5, 6 | 515 | |||
U08372.1 | - | 4, 5 | 562 | |||
U09362.1 | - | 5, 6 | 463 |