Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ate1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029895.3 | NP_001025066.1 | 9 | 1496 | ||
NM_001136054.2 | NP_001129526.1 | 9 | 1228 | |||
NM_001271343.1 | NP_001258272.1 | 9 | 1496 | |||
NM_013799.3 | NP_038827.2 | 9 | 1228 | |||
GenBank mRNA | AK077471.1 | - | 5 | 647 | ||
AK148442.1 | - | 9 | 1196 | |||
AK167591.1 | - | 9 | 1188 | |||
BC042601.1 | - | 9 | 1187 | 4503326 | ||
BC050948.1 | - | 9 | 1178 | 6826691 | ||
DQ093639.1 | - | 9 | 1088 | |||
DQ093640.1 | - | 9 | 1088 |