Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007497.3 | NP_031523.3 | 6 | 881 | ||
GenBank mRNA | AK010428.1 | - | 6 | 762 | ||
AK144511.1 | - | 6 | 924 | |||
BC006871.1 | - | 6 | 827 | 2646620 | ||
M63725.1 | - | 5 | 671 |