Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007497.3 | NP_031523.3 | 3 | 364 | ||
GenBank mRNA | AK010428.1 | - | 3 | 245 | ||
AK144511.1 | - | 3 | 406 | |||
BC006871.1 | - | 3 | 310 | 2646620 | ||
BY733806.1 | - | 3 | 403 | |||
M63725.1 | - | 2 | 154 |