Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287180.1 | NP_001274109.1 | 3 | 408 | ||
NM_009716.3 | NP_033846.2 | 2 | 691 | |||
GenBank mRNA | AA003141.1 | - | 3 | 407 | ||
AK003001.1 | - | 2 | 692 | |||
AK138657.1 | - | 2 | 376 | |||
AK144777.1 | - | 2 | 377 | |||
AK146193.1 | - | 2 | 379 | |||
AK150231.1 | - | 2 | 378 | |||
AK155254.1 | - | 2 | 377 | |||
AK156298.1 | - | 2 | 371 | |||
AK157839.1 | - | 2 | 346 | |||
BC031542.1 | - | 1 | 873 | |||
BC085169.1 | - | 2 | 582 | 30551592 | ||
L13791.1 | - | 1 | 196 | |||
M94087.1 | - | 2 | 358 | |||
X61507.1 | - | 2 | 368 |