Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neurod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007501.4 | NP_031527.1 | 2 | 1038 | ||
GenBank mRNA | AK012183.1 | - | 2 | 1025 | ||
BC042639.1 | - | 2 | 987 | |||
BC054391.1 | - | 2 | 1038 | 6491248 | ||
D85845.1 | - | 1 | 945 |