Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144900.2 | NP_659149.1 | 4 | 649 | ||
GenBank mRNA | AK088000.1 | - | 4 | 610 | ||
AK137001.1 | - | 4 | 608 | |||
AK159279.1 | - | 4 | 606 | |||
AK159306.1 | - | 4 | 606 | |||
AK181228.1 | - | 3 | 319 | |||
AK191920.1 | - | 1 | 77 | |||
AK206258.1 | - | 1 | 148 | |||
AK207531.1 | - | 1 | 77 | |||
BC010319.1 | - | 2 | 234 | |||
BC021496.1 | - | 4 | 599 | 5346159 | ||
BC023794.1 | - | 5 | 544 | 5351943 | ||
BC023964.1 | - | 4 | 589 | |||
BC025618.1 | - | 4 | 608 | 5320976 | ||
BC025627.1 | - | 4 | 599 | 5320513 | ||
BC025811.1 | - | 4 | 586 | 5096872 | ||
BC032187.1 | - | 4 | 649 | 5321268 | ||
BC033435.1 | - | 4 | 545 | 4950746 | ||
BC033471.1 | - | 4 | 589 | 4954119 | ||
BC034177.1 | - | 3 | 354 | |||
BC042435.1 | - | 4 | 599 | 4951288 |