Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007504.2 | NP_031530.2 | 4 | 416 | ||
GenBank mRNA | AY081946.1 | - | 4 | 447 | ||
BC036292.1 | - | 4 | 416 | 4191741 |