Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000849.4 | NP_000840.2 | 6 | 617 | ||
NR_024537.1 | 5 | 541 | ||||
GenBank mRNA | AB209816.1 | - | 5 | 326 | ||
AK304300.1 | - | 4 | 269 | |||
AK313444.1 | - | 5 | 335 | |||
BC000088.2 | - | 5 | 322 | 3509784 | ||
BC008790.2 | - | 6 | 617 | 3630923 | ||
BT019945.1 | - | 5 | 307 | |||
CR542108.1 | - | 5 | 307 | |||
EF363096.1 | - | 5 | 324 | |||
J05459.1 | - | 5 | 324 |