Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007505.2 | NP_031531.1 | 10 | 1687 | ||
GenBank mRNA | AK043976.1 | - | 10 | 1455 | ||
AK076572.1 | - | 10 | 1455 | |||
AK098155.1 | - | 10 | 1465 | |||
AK146797.1 | - | 10 | 1353 | |||
AK150426.1 | - | 10 | 1458 | |||
AK150843.1 | - | 10 | 1455 | |||
AK151004.1 | - | 10 | 1455 | |||
AK151128.1 | - | 10 | 1464 | |||
AK151224.1 | - | 10 | 1462 | |||
AK151920.1 | - | 10 | 1459 | |||
AK152054.1 | - | 10 | 1466 | |||
AK152890.1 | - | 10 | 1458 | |||
AK159491.1 | - | 10 | 1455 | |||
AK159540.1 | - | 10 | 1455 | |||
AK159758.1 | - | 10 | 1458 | |||
AK160043.1 | - | 10 | 1465 | |||
AK164110.1 | - | 10 | 1458 | |||
AK164193.1 | - | 8 | 1161 | |||
AK166709.1 | - | 10 | 1458 | |||
AK166812.1 | - | 10 | 1471 | |||
AK167159.1 | - | 10 | 1455 | |||
AK167863.1 | - | 10 | 1435 | |||
AK168198.1 | - | 10 | 1465 | |||
AK168369.1 | - | 10 | 1457 | |||
AK168617.1 | - | 10 | 1455 | |||
AK168879.1 | - | 10 | 1455 | |||
AK168890.1 | - | 10 | 1455 | |||
AK168932.1 | - | 10 | 1455 | |||
AK169080.1 | - | 10 | 1458 | |||
AK169084.1 | - | 10 | 1458 | |||
AK169105.1 | - | 10 | 1455 | |||
AK169142.1 | - | 10 | 1500 | |||
AK169300.1 | - | 10 | 1455 | |||
AK169308.1 | - | 10 | 1460 | |||
AK169414.1 | - | 10 | 1458 | |||
AK192928.1 | - | 1 | 84 | |||
AK200623.1 | - | 84 | ||||
BC014854.1 | - | 10 | 1442 | 4500893 | ||
BC020417.1 | - | 10 | 1442 | |||
BC055892.1 | - | 10 | 1892 | |||
L01062.1 | - | 10 | 1442 |