Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007508.5 | NP_031534.2 | 2 | 116 | ||
GenBank mRNA | AK140873.1 | - | 2 | 92 | ||
AK149833.1 | - | 2 | 131 | |||
AK150742.1 | - | 2 | 102 | |||
AK153403.1 | - | 2 | 117 | |||
AK154869.1 | - | 2 | 117 | |||
AK160792.1 | - | 2 | 131 | |||
AK166857.1 | - | 2 | 106 | |||
AK170721.1 | - | 2 | 117 | |||
AK210420.1 | - | 1 | 65 | |||
AK217192.1 | - | 1 | 65 | |||
AK217211.1 | - | 1 | 65 | |||
BC038392.1 | - | 2 | 94 | 2651677 | ||
BC050135.1 | - | 1 | 59 |