Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289445.1 | NP_001276374.1 | 8 | 939 | ||
NM_001289446.1 | NP_001276375.1 | 8 | 669 | |||
NM_015731.3 | NP_056546.2 | 8 | 739 | |||
GenBank mRNA | AK089064.1 | - | 8 | 845 | ||
BC006949.1 | - | 6 | 441 | |||
BC021814.1 | - | 8 | 739 | |||
BC075718.1 | - | 8 | 867 | |||
BC094551.1 | - | 8 | 709 | 6813369 |