Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atpif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF002718.1 | - | 2, 3 | 217 | ||
AK008346.1 | - | 2, 3 | 215 | |||
AK152389.1 | - | 3, 4 | 1232 | |||
AV462016.1 | - | 2, 3 | 278 | |||
BC012680.1 | - | 2, 3 | 276 | 4217188 | ||
BU055998.1 | - | 2, 3 | 203 | |||
CB848021.2 | - | 2, 3 | 240 |