Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atpif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF002718.1 | - | 3 | 263 | ||
AK008346.1 | - | 3 | 261 | |||
AK152389.1 | - | 4 | 1278 | |||
AV462016.1 | - | 3 | 324 | |||
BC012680.1 | - | 3 | 322 | 4217188 | ||
BU055998.1 | - | 3 | 250 | |||
CB848021.2 | - | 3 | 286 |