Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080843.3 | NP_001074312.1 | 5 | 829 | ||
XM_005261715.4 | XP_005261772.1 | 5 | 807 | |||
GenBank mRNA | BC002415.1 | - | 5 | 834 | 3164017 | |
BC071700.1 | - | 5 | 801 | 4932043 | ||
BC146936.1 | - | 5 | 818 | 9056864 |
Gene Symbol: GSTT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000854.4 | NP_000845.2 | 5 | 949 | ||
NM_001302670.1 | NP_001289599.1 | 5 | 907 | |||
NR_126442.1 | 6 | 941 | ||||
NR_126445.1 | 5 | 949 | ||||
NR_126446.1 | 6 | 1011 | ||||
GenBank mRNA | BC146929.1 | - | 5 | 818 | 9056857 | |
BC146940.1 | - | 5 | 919 | 9056868 | ||
BC146943.1 | - | 5 | 919 | 9056871 | ||
CD013993.1 | - | 6 | 819 | |||
CR456374.1 | - | 5 | 759 | |||
CR456500.1 | - | 5 | 799 | |||
L38503.1 | - | 5 | 779 |