Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301424.1 | NP_001288353.1 | 8 | 1256 | ||
NM_007513.4 | NP_031539.3 | 8 | 1372 | |||
GenBank mRNA | AK137512.1 | - | 1 | 2119 | ||
AK139342.1 | - | 8 | 1348 | |||
AK147991.1 | - | 8 | 1281 | |||
AK163202.1 | - | 8 | 1256 | |||
BC068234.1 | - | 8 | 1263 | |||
BC145779.1 | - | 7 | 1177 | 40131082 | ||
BC145781.1 | - | 7 | 1177 | 40131084 | ||
JN226408.1 | - | 6 | 1092 | |||
JN226410.1 | - | 6 | 1092 | |||
M26687.1 | - | 8 | 1290 |