Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001312660.1 | NP_001299589.1 | 3, 4 | 529 | ||
NM_145870.2 | NP_665877.1 | 2, 3 | 343 | |||
NM_145871.2 | NP_665878.2 | 2, 3 | 343 | |||
XM_011536670.2 | XP_011534972.1 | 3, 4 | 585 | |||
XM_011536671.2 | XP_011534973.1 | 2, 3 | 253 | |||
XM_017021243.1 | XP_016876732.1 | 2, 3 | 255 | |||
GenBank mRNA | AJ001838.1 | - | 2, 3 | 164 | ||
AK309054.1 | - | 2, 3 | 191 | |||
AK315154.1 | - | 3, 4 | 420 | |||
AK315609.1 | - | 2, 3 | 187 | |||
BC001453.1 | - | 2, 3 | 219 | 3139094 | ||
BE958109.1 | - | 150 | ||||
BG330365.1 | - | 2, 3 | 205 | |||
BG772450.1 | - | 2, 3 | 350 | |||
BI559916.1 | - | 3, 4 | 438 | |||
BI829992.1 | - | 3, 4 | 409 | |||
CR456987.1 | - | 1, 2 | 61 | |||
U86529.1 | - | 2, 3 | 164 |