Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Axin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015732.4 | NP_056547.3 | 3 | 1188 | ||
GenBank mRNA | AF073788.1 | - | 3 | 1053 | ||
AF205889.1 | - | 2 | 893 | |||
AK049192.1 | - | 3 | 1132 | |||
AK142333.1 | - | 3 | 1190 | |||
AK197385.1 | - | 2 | 136 | |||
BC057338.1 | - | 3 | 1171 | 6827741 |