Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007528.3 | NP_031554.1 | 4 | 617 | ||
GenBank mRNA | AB011665.1 | - | 3 | 540 | ||
AK156733.1 | - | 4 | 579 | |||
BC090998.1 | - | 4 | 577 | 6750884 |