Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007528.3 | NP_031554.1 | 7 | 1184 | ||
GenBank mRNA | AB011665.1 | - | 6 | 1107 | ||
AK032647.1 | - | 2 | 175 | |||
AK156733.1 | - | 7 | 1146 | |||
BC090998.1 | - | 7 | 1144 | 6750884 |