Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bckdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199195.1 | NP_954665.1 | 3 | 264 | ||
GenBank mRNA | AK089921.1 | - | 3 | 342 | ||
AK156264.1 | - | 3 | 339 | |||
AV455466.1 | - | 3 | 352 | |||
BC064099.1 | - | 3 | 264 | 1246285 | ||
L16992.1 | - | 3 | 336 |