Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bckdk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009739.3 | NP_033869.1 | 11 | 1260 | ||
GenBank mRNA | AF043070.1 | - | 10 | 1101 | ||
AK075602.1 | - | 11 | 1186 | |||
AK145479.1 | - | 11 | 1231 | |||
AK150607.1 | - | 11 | 1252 | |||
AK150723.1 | - | 11 | 1252 | |||
AK167519.1 | - | 11 | 1252 | |||
AK193909.1 | - | 1 | 50 | |||
AK207870.1 | - | 2 | 105 | |||
AK218004.1 | - | 2 | 105 | |||
BC004077.1 | - | 8 | 688 | |||
BC046595.1 | - | 11 | 1230 | 6313106 |