Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009740.2 | NP_033870.1 | 3 | 648 | ||
GenBank mRNA | AF057701.1 | - | 3 | 532 | ||
AF100339.1 | - | 3 | 540 | |||
AF127387.1 | - | 3 | 451 | |||
AF134396.1 | - | 3 | 532 | |||
AJ006289.1 | - | 3 | 529 | |||
AK076082.1 | - | 3 | 560 | |||
AK140179.1 | - | 3 | 520 | |||
AK146236.1 | - | 3 | 554 | |||
AK150883.1 | - | 3 | 549 | |||
AK152563.1 | - | 3 | 549 | |||
AK152847.1 | - | 3 | 545 | |||
AK156890.1 | - | 3 | 602 | |||
AK168084.1 | - | 3 | 554 | |||
AK169126.1 | - | 3 | 555 | |||
AK172158.1 | - | 3 | 559 | |||
AK207365.1 | - | 2 | 140 | |||
BC003703.1 | - | 3 | 567 | |||
BC024379.1 | - | 3 | 526 | 4976147 | ||
BC172007.1 | - | 3 | 532 | 9054293 | ||
BC172009.1 | - | 3 | 532 | 9054295 | ||
BU707261.1 | - | 3 | 648 |