Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278940.1 | NP_001265869.1 | 10 | 1556 | ||
NM_015859.3 | NP_056943.1 | 9 | 1489 | |||
NM_201595.2 | NP_963889.1 | 9 | 1109 | |||
GenBank mRNA | BC107155.2 | - | 9 | 1224 | 40032646 | |
BC107156.1 | - | 9 | 1226 | 40032650 | ||
BX493028.1 | - | 3 | 167 | |||
D14886.1 | - | 9 | 1109 | |||
D14887.1 | - | 9 | 1459 | |||
X75383.1 | - | 9 | 1196 | |||
X77225.1 | - | 9 | 1055 |