Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320929.1 | NP_001307858.1 | 2, 3 | 252 | ||
NM_001320930.1 | NP_001307859.1 | 3, 4 | 476 | |||
NM_004492.2 | NP_004483.1 | 2, 3 | 254 | |||
GenBank mRNA | AK312015.1 | - | 2, 3 | 254 | ||
BC000287.2 | - | 2, 3 | 216 | 2819215 | ||
BC001919.1 | - | 2, 3 | 216 | 2819215 | ||
BT007362.1 | - | 1, 2 | 72 | |||
CB156109.1 | - | 3, 4 | 418 | |||
CR542192.1 | - | 1, 2 | 72 | |||
HY002503.1 | - | 2, 3 | 239 | |||
HY036972.1 | - | 2, 3 | 252 | |||
U14193.1 | - | 2, 3 | 213 | |||
U21242.1 | - | 2, 3 | 182 | |||
X81713.1 | - | 2, 3 | 171 |