Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009745.2 | NP_033875.2 | 3 | 315 | ||
GenBank mRNA | AJ011145.1 | - | 3 | 195 | ||
AK131643.1 | - | 3 | 211 | |||
AK155076.1 | - | 3 | 197 | |||
BC011471.1 | - | 3 | 292 | 3660754 | ||
BY087167.1 | - | 3 | 315 |