Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009752.2 | NP_033882.1 | 13 | 1365 | ||
GenBank mRNA | AK034966.1 | - | 1257 | |||
AK042069.1 | - | 13 | 1335 | |||
AK048460.1 | - | 13 | 1296 | |||
AK076517.1 | - | 13 | 1336 | |||
AK153218.1 | - | 13 | 1347 | |||
AK166473.1 | - | 13 | 1351 | |||
AK171593.1 | - | 13 | 1368 | |||
AK204903.1 | - | 5 | 343 | |||
AK217231.1 | - | 4 | 311 | |||
BC003998.1 | - | 11 | 936 | |||
BC028875.1 | - | 13 | 1285 | 4240285 | ||
M57734.1 | - | 13 | 1324 |