Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009752.2 | NP_033882.1 | 13 | 1432 | ||
GenBank mRNA | AK034966.1 | - | 1324 | |||
AK042069.1 | - | 13 | 1402 | |||
AK048460.1 | - | 13 | 1363 | |||
AK076517.1 | - | 13 | 1403 | |||
AK153218.1 | - | 13 | 1414 | |||
AK171593.1 | - | 13 | 1435 | |||
M57734.1 | - | 13 | 1391 |