Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bhmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016668.3 | NP_057877.1 | 3 | 486 | ||
GenBank mRNA | AF033381.1 | - | 3 | 229 | ||
AK077108.1 | - | 3 | 293 | |||
AK145668.1 | - | 3 | 328 | |||
AK149431.1 | - | 3 | 295 | |||
AK149457.1 | - | 3 | 293 | |||
AK166754.1 | - | 3 | 295 | |||
AK166818.1 | - | 3 | 294 | |||
BC029020.1 | - | 1 | 605 | |||
BC037004.1 | - | 3 | 288 | 5100429 | ||
BC093510.1 | - | 3 | 261 | 5101184 | ||
BC110307.1 | - | 3 | 689 | 5310035 |