Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTBN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003128.2 | NP_003119.2 | 25, 26 | 5458 | ||
NM_178313.2 | NP_842565.2 | 24, 25 | 5555 | |||
XM_005264517.2 | XP_005264574.1 | 25, 26 | 5367 | |||
XM_005264518.1 | XP_005264575.1 | 24, 25 | 6213 | |||
XM_006712087.2 | XP_006712150.1 | 25, 26 | 5331 | |||
XM_017004779.1 | XP_016860268.1 | 25, 26 | 5474 | |||
XM_017004780.1 | XP_016860269.1 | 25, 26 | 5428 | |||
XM_017004781.1 | XP_016860270.1 | 25, 26 | 5429 | |||
GenBank mRNA | AB209748.1 | - | 25, 26 | 5336 | ||
AB371586.1 | - | 25, 26 | 5448 | |||
AF327441.1 | - | 24, 25 | 5555 | |||
BC137282.1 | - | 24, 25 | 5465 | 9021279 | ||
BC137283.1 | - | 24, 25 | 5465 | 9021280 | ||
EU794662.1 | - | 25, 26 | 5458 |