Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007552.4 | NP_031578.2 | 7, 8 | 947 | ||
GenBank mRNA | BC053708.1 | - | 7, 8 | 573 | 30074540 | |
BC056384.1 | - | 6, 7 | 477 | 6854104 | ||
DQ337175.1 | - | 6, 7 | 476 | |||
M64279.1 | - | 7, 8 | 947 |