Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256019.1 | NP_001242948.1 | 1 | 772 | ||
NM_007558.3 | NP_031584.1 | 1 | 772 | |||
GenBank mRNA | AK157978.1 | - | 1 | 434 | ||
BC052168.1 | - | 1 | 770 | 6334507 | ||
M97017.1 | - | 1 | 320 |