Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bnc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007562.2 | NP_031588.2 | 5 | 2831 | ||
GenBank mRNA | AK087567.1 | - | 5 | 2826 | ||
BC127226.1 | - | 1 | 498 | |||
U88064.1 | - | 5 | 2778 |