Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bnip3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009761.3 | NP_033891.1 | 3 | 406 | ||
GenBank mRNA | AF067395.1 | - | 3 | 412 | ||
AK004667.1 | - | 3 | 353 | |||
AK007920.1 | - | 3 | 389 | |||
AK013467.1 | - | 3 | 388 | |||
AK018668.1 | - | 3 | 358 | |||
AK139950.1 | - | 3 | 350 | |||
AK155040.1 | - | 3 | 377 | |||
AK168482.1 | - | 3 | 407 | |||
AK169391.1 | - | 3 | 407 | |||
AY057069.1 | - | 3 | 385 | |||
BC049081.1 | - | 3 | 319 | |||
BC085237.1 | - | 3 | 356 | 5362586 | ||
BU560379.1 | - | 3 | 409 |