Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bnip3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009761.3 | NP_033891.1 | 1 | 143 | ||
GenBank mRNA | AF067395.1 | - | 1 | 149 | ||
AK004667.1 | - | 1 | 90 | |||
AK007920.1 | - | 1 | 126 | |||
AK013467.1 | - | 1 | 125 | |||
AK018668.1 | - | 1 | 95 | |||
AK139950.1 | - | 1 | 87 | |||
AK155040.1 | - | 1 | 114 | |||
AK168482.1 | - | 1 | 144 | |||
AK169391.1 | - | 1 | 144 | |||
AY057069.1 | - | 1 | 122 | |||
BC049081.1 | - | 1 | 56 | |||
BC085237.1 | - | 1 | 93 | 5362586 | ||
BU560379.1 | - | 1 | 146 |